MABS1256
Anti-GRK2 Antibody, clone E23/8
clone E23/8, from mouse
Synonym(s):
Beta-adrenergic receptor kinase 1, Beta-ARK-1, G-protein-coupled receptor kinase 2
About This Item
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
E23/8, monoclonal
species reactivity
monkey, bovine, rat, human
species reactivity (predicted by homology)
mouse (based on 100% sequence homology)
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... GRK2(156)
mouse ... Grk2(282682) , Grk2(110355)
rat ... Grk2(25238)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected delta opioid receptor agonists- (DPDPE & DADLE) induced interaction between co-expressed bovine GRK2 and HA-tagged full-length murine delta opioid receptor (DOR), but not HA-mDOR lacking C-terminal end 31 residues, in HEK293 transfectants by Western blotting after anti-HA IP. Naloxone co-treatment abolished DPDPE effect (Li, J. et al. (2003). J. Biol. Chem. 278(32):30219-30226).
Western Blotting Analysis: A representative lot detected the expression of virally transfected full-length bovine GRK2 and C-terminal fragment (GRK2ct) in rabbit smooth muscle cells (Peppel, K., et al. (2000). Circulation. 102(7):793-799).
ELISA Analysis: Clone E23/8 was biotinylated and employed as the detection antibody for the detection of recombinant bovine GRK2 as well as endogenous GRK2 in human CCR5-expressing rat basophilic leukemia RBL-2H3 cells (RBL-CCR5), COS cells, and human leukocytes (lymphocytes, monocytes, granulocytes) by sandwich ELISA (Oppermann, M., et al. (1999). J. Biol. Chem. 274:8875 -8885).
Quality
Western Blotting Analysis: A 1:250 dilution of this antibody detected the exogenously expressed bovine GRK2 in lysate from transfected HEK293 cells.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service