Skip to Content
Merck
All Photos(1)

Key Documents

MABN543

Sigma-Aldrich

Anti-Gs protein Antibody, alpha subunit, clone N192/12

clone N192/12, from mouse

Synonym(s):

Guanine nucleotide-binding protein G(s) subunit alpha isoforms short, Adenylate cyclase-stimulating G alpha protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

N192/12, monoclonal

species reactivity

rat, human

species reactivity (predicted by homology)

bovine (immunogen homology)

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GNAS(2778)

Related Categories

General description

The heterotrimeric guanine nucleotide-binding regulatory proteins (G proteins) mediate signaling from a large number of G-protein coupled receptors (GPCR) to a variety of intracellular effectors. G-proteins are composed of a 36-52 kDa α-subunit, a 35-36 kDa β-subunit, and an 8-10 kDa γ-subunit. They are classified based on the nature of their α-subunit. Gs is ubiquitously expressed and couples receptors to adenylyl cyclase and promotes intracellular cAMP generation. Mutations in Gs-α can lead to constitutive activation of receptors leading to development of endocrine tumors, fibrous dysplasia of bone, and McCune-Albright syndrome. In brain tissue Gs-α activates adenyl cyclase in direct response to stimulation of several neurotransmitter receptors and facilitates depolarization-induced neurotransmitter release for synaptic transmission. Also, it is shown that Gs-stimulated adenylyl cyclase activity is significantly reduced in the Alzheimer′s disease brain tissue.

Specificity

Other homologies: Mouse (99% sequence homology).

Immunogen

Recombinant protein corresponding to bovine GS protein, alpha subunit.

Application

Anti-Gs protein Antibody, alpha subunit, clone N192/12 is a highly specific mouse monoclonal antibody & that targets Gsα & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected GS protein, alpha subunit in 10 µg of SH-SY5Y cell lysate.

Quality

Evaluated by Western Blotting in rat brain membrane tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected GS protein, alpha subunit in 10 µg of rat brain membrane tissue lysate.

Target description

~45 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain membrane tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guilherme Ruiz Leonardi et al.
Journal of thrombosis and haemostasis : JTH, 20(7), 1699-1711 (2022-04-09)
Thrombotic antiphospholipid syndrome (t-PAPS) is characterized by arterial, venous, or microvascular occlusions, which are explained, in part, by the presence of antiphospholipid (aPL) antibodies. Although there is much evidence indicating that isolated aPL antibodies increase the activity of platelets obtained
Jessica J Chen et al.
Biophysical journal, 118(4), 826-835 (2019-09-25)
S-palmitoylation is a reversible posttranslational modification that plays an important role in regulating protein localization, trafficking, and stability. Recent studies have shown that some proteins undergo extremely rapid palmitoylation/depalmitoylation cycles after cellular stimulation supporting a direct signaling role for this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service