Skip to Content
Merck
All Photos(1)

Documents

MABF449

Sigma-Aldrich

Anti-CD45RA Antibody (human), PerCP-Cy5.5, clone HI100

clone HI100, from mouse

Synonym(s):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, T200, CD antigen CD45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

PerCP-Cy5.5

antibody form

purified antibody

antibody product type

primary antibodies

clone

HI100, monoclonal

species reactivity

human

packaging

pkg of 5 μL (0.06 μg/test)

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPRC(5788)

Related Categories

General description

The HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RB, CD45RO and others.The HI100 antibody is widely used as a marker for human CD45RA expression on naïve and activated T cells, B cells, and monocytes.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This mouse monoclonal antibody Anti-CD45RA (human), PerCP-Cy5.5, clone HI100 validated for use in western blotting, IHC & Flow Cytometry for the detection of CD45RA.

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.06 µg of this antibody detected CD45RA in one million human peripheral blood lymphocytes.

Linkage

Replaces: 04-1102

Physical form

Purified Mouse Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service