Skip to Content
Merck
All Photos(2)

Key Documents

MABE1784

Sigma-Aldrich

Anti-NCoA-2 Antibody, clone 3Ti-3C11

Synonym(s):

Class E basic helix-loop-helix protein 75 (bHLHe75), NCoA-2, SRC2, Transcriptional intermediary factor 2 (hTIF2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3Ti-3C11, monoclonal

mol wt

calculated mol wt 159 kDa
observed mol wt ~85 kDa

purified by

using protein G

species reactivity

mouse, human

species reactivity (predicted by homology)

feline, horse

packaging

antibody small pack of 100

technique(s)

ChIP: suitable
ELISA: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... NCOA2(10499)

Specificity

Clone 3Ti-3C11 mouse monoclonal antibody that detects NCoA-2. It targets an epitope within 246 amino acids from the internal part of the protein.

Immunogen

His-tagged, recombinant, human NCoA-2 protein fragment (aa 624-1287).

Application

Quality Control Testing

Evaluated by Western Blotting with His-tagged, recombinant fragment of Human NCoA-2.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected His-tagged, recombinant fragment (aa 624-1287) of Human NCoA-2.

Tested Applications

ELISA Analysis: A representative lot detected His-tagged, recombinant fragment (aa 624-1287) of Human NCoA-2.

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected NCoA-2 in Chromatin Immunoprecipitation application. (Rambaud, J., et al. (2009). J Biol Chem. 284(21):14147-56; Duteil, D., et al. (2010). Cell Metab. 12(5):496-508; Ganti, K.P., et al. (2017). Proc Natl Acad Sci USA. 114(6):E951-E960).

Western Blotting Analysis: A representative lot detected NCoA-2 in Western Blotting application. (Rambaud, J., et al. (2009). J Biol Chem. 284(21):14147-56; Duteil, D., et al. (2010). Cell Metab. 12(5):496-508).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Nuclear receptor coactivator 2 (UniProt: Q15596; also known as NCoA-2, Transcriptional intermediary factor 2, hTIF2, Class E basic helix-loop-helix protein 75 (bHLHe75)) is encoded by the NCOA2 (also known as BHLHE75, SRC2, TIF2) gene (Gene ID: 10499) in human. Nuclear receptor coactivator 2 (NCoA-2) functions as a transcriptional coactivator in various cellular processes. It belongs to the p160/steroid receptor coactivator (SRC) family of proteins. NCoA-2 plays a crucial role in the regulation of gene expression by interacting with nuclear receptors and other transcription factors. It possesses multiple functional domains, including a nuclear receptor interaction domain (NRID), a histone acetyltransferase (HAT) domain, and several transcription activation domains (ADs). NCoA-2 acts as a bridge between transcription factors and the transcriptional machinery, facilitating the recruitment of coactivators and chromatin remodeling complexes to target gene promoters. This coactivator function enables NCoA-2 to modulate diverse biological processes such as development, metabolism, cell proliferation, and differentiation. Dysregulation of NCoA-2 has been implicated in various diseases, including cancer, where it can promote tumor growth and progression.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service