EZFITMIC06
EZ-Fit® Manifold
6-place, for Microfil® funnels and membranes
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
material
stainless steel
Sterilization Compatibility
autoclavable compatible
packaging
pkg of 1 unit
manufacturer/tradename
EZ-Fit®
input
sample type cosmetics
sample type beer
sample type pharmaceutical(s)
sample type bottled water
application(s)
bioburden testing
cosmetics
environmental
food and beverages
water monitoring
shipped in
ambient
Related Categories
General description
EZ-Fit® Manifold helps to optimize the workflow and take control of the contamination risks. Every single component of the manifold can be removed for easy cleaning and autoclaving, thus preventing the formation of biofilm. The 2-way valve design prevents the user from accidentally touching the filtration head while opening or closing the valve. A low height design increases comfort when used in laminar flow hoods. The new Microfil® head design improves ease of use, reduces maintenance, and limits the risk of contamination. A continuous rim allows easy and safe membrane transfer with forceps, reducing the risk of touching the membrane filtration area. The porous filtration support can be removed simply by pressing down anywhere around its edge.
Application
The EZ-Fit® filtration Manifold is used for universal laboratory filtrations and supports the simultaneous filtration of six test samples. The manifold is suitable to be used with Microfil® filtration units.
Legal Information
EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
configured for
Product No.
Description
Pricing
recommended
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service