Skip to Content
Merck
All Photos(1)

Documents

DX2510

Supelco

Drierite non-Indicating

8 Mesh

Synonym(s):

Drierite non-Indicating, desiccant

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaO4S
CAS Number:
Molecular Weight:
136.14
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-900-3

form

granular

size

8 mesh

color

white

particle size

8 mesh

mp

1450 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

OSGAYBCDTDRGGQ-UHFFFAOYSA-L

Application

8 Mesh

Analysis Note

Description: White irregular granules

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service