CXXC finger protein 1 (CFP1) is part of the Setd1A and Setd1B methyltransferase compounds. Research shows that CFP1 also interacts with the cytosine methylation mechanism. CFP1 is necessary for development of mammals and is a key regulator of chromatin structure. Embryonic stem cells that are missing CFP1 are more susceptible to DNA-damaging agents and tend to build up more DNA damage. CFP1 has been shown to also be a regulator of DNA repair.
Immunogen
GST-tagged recombinant protein corresponding to human CFP1.
Application
Use Anti-CFP1 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect CFP1 also known as CXXC finger 1 (PHD domain), cpG-binding protein.
Quality
Evaluated by Western Blot in HeLa nuclear extract.
Western Blot Analysis: 1:1,000 dilution of this antibody detected CFP1 on 10 µg of HeLa nuclear extract.
Target description
~78 kDa observed. A cross-reacting band observed at ~110 kDa
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Transposable elements (TEs) are mobile genetic elements that make up a large fraction of mammalian genomes. While select TEs have been co-opted in host genomes to have function, the majority of these elements are epigenetically silenced by DNA methylation in
PR domain-containing protein 9 (PRDM9) is a major regulator of the localization of meiotic recombination hotspots in the human and mouse genomes. This role involves its DNA-binding domain, which is composed of a tandem array of zinc fingers, and PRDM9-dependent
MAF1 represses Pol III-mediated transcription by interfering with TFIIIB and Pol III. Herein, we found that MAF1 knockdown induced CDKN1A transcription and chromatin looping concurrently with Pol III recruitment. Simultaneous knockdown of MAF1 with Pol III or BRF1 (subunit of
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.