AB9304
Anti-NAP-22 Antibody, complete sequence
Chemicon®, from rabbit
Synonym(s):
Neuronal acidic protein 22, CAP-23, BASP1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
biological source
rabbit
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
rat
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
rat ... Basp1(64160)
Specificity
Recognizes NAP-22 [Neuronal acidic protein 22, CAP-23, BASP1].
Immunogen
Epitope: Complete sequence
Recombinant full length rat NAP-22.
Application
Immunoblotting: 1:1,000 using rat brain extract (colormetric; higher dilutions may be used with ECL). The antibody reacts with the 55 kDa NAP-22 protein. The suggested dilution buffer is TBS containing 0.05% Tween-20. Suggested blocking buffer is 5% non-fat dry milk. Suggested transfer membrane is PVDF-Immobilon. Suggested gel percentage is 10%. Overnight incubation with the antibody is recommended.
Optimal working dilutions must be determined by the end user.
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Neuroscience
Research Sub Category
Developmental Neuroscience
Neuroregenerative Medicine
Developmental Neuroscience
Neuroregenerative Medicine
This Anti-NAP-22 Antibody, complete sequence is validated for use in WB for the detection of NAP-22.
Physical form
Format: Purified
Semi-purified immunoglobulin. Liquid in TBS containing 0.05% sodium azide.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.
Analysis Note
Control
POSITIVE CONTROL: Neonatal rat brain extract. NAP-22 is heat and acid resistant. Heat or acid-treated supernatants from crude tissue or cell homogenates are good sources to analyze NAP-22 content by immunoblot.
POSITIVE CONTROL: Neonatal rat brain extract. NAP-22 is heat and acid resistant. Heat or acid-treated supernatants from crude tissue or cell homogenates are good sources to analyze NAP-22 content by immunoblot.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service