Skip to Content
Merck
All Photos(1)

Documents

8.52259

Sigma-Aldrich

(9-Fluorenylmethyl) chloroformate

Novabiochem®

Synonym(s):

(9-Fluorenylmethyl) chloroformate, (9-Fluorenylmethyl) chloroformate, Chloroformic acid 9-fluorenmethyl ester, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO2
CAS Number:
Molecular Weight:
258.70
MDL number:
UNSPSC Code:
12352107
EC Index Number:
249-313-6
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥98.0% (HPLC)

form

powder

manufacturer/tradename

Novabiochem®

mp

60-63 °C

application(s)

peptide synthesis

storage temp.

2-8°C

InChI

1S/C15H11ClO2/c16-15(17)18-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14H,9H2

InChI key

IRXSLJNXXZKURP-UHFFFAOYSA-N

General description

Reagent for introduction of the Fmoc protecting group [1].

Literature references

[1] L.A.Carpino et al. (1987) Acc. Chem. Res., 20, 401.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 60 °C
Melting range (upper value): ≤ 64 °C
Identity (IR): passes test

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service