Skip to Content
Merck
All Photos(1)

Documents

8.41070

Sigma-Aldrich

Pentachloropyridine

for synthesis

Synonym(s):

Pentachloropyridine, Perchloropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5Cl5N
CAS Number:
Molecular Weight:
251.33
UNSPSC Code:
12352101
EC Index Number:
218-535-5
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

powder

potency

435 mg/kg LD50, oral (Rat)

mp

124-126 °C

storage temp.

2-30°C

InChI

1S/C5Cl5N/c6-1-2(7)4(9)11-5(10)3(1)8

InChI key

DNDPLEAVNVOOQZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 123 °C
Melting range (upper value): ≤ 126 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service