Skip to Content
Merck
All Photos(2)

Key Documents

8.21685

Sigma-Aldrich

Benzyltriphenylphosphonium chloride

for synthesis

Synonym(s):

Benzyltriphenylphosphonium chloride, Triphenylbenzylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2(C6H5)3PCl
CAS Number:
Molecular Weight:
388.87
MDL number:
UNSPSC Code:
12352107
EC Index Number:
214-154-3
Pricing and availability is not currently available.

vapor pressure

0.1 hPa

potency

43 mg/kg LD50, oral (Rat)

mp

337 °C

density

1.18 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

2-25°C

InChI

1S/C25H22P.ClH/c1-5-13-22(14-6-1)21-26(23-15-7-2-8-16-23,24-17-9-3-10-18-24)25-19-11-4-12-20-25;/h1-20H,21H2;1H/q+1;/p-1

InChI key

USFRYJRPHFMVBZ-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service