Skip to Content
Merck
All Photos(1)

Documents

8.20619

Sigma-Aldrich

2-Heptanol

for synthesis

Synonym(s):

2-Heptanol, Methyl pentyl carbinol, Methyl amyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH3
CAS Number:
Molecular Weight:
116.20
MDL number:
UNSPSC Code:
12352104
EC Index Number:
208-844-3
NACRES:
NA.22

vapor pressure

1.6 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

potency

2580 mg/kg LD50, oral (Rat)
1460 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9 % (v/v)

transition temp

flash point 59 °C

solubility

3.27 g/L

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3

InChI key

CETWDUZRCINIHU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.816 - 0.818
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F

Flash Point(C)

59 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service