Skip to Content
Merck
All Photos(1)

Documents

8.20515

Sigma-Aldrich

N,N-Dimethyl-4-nitrosoaniline

for synthesis

Synonym(s):

N,N-Dimethyl-4-nitrosoaniline, 4-Nitroso-N,N-dimethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(NO)C6H4N(CH3)2
CAS Number:
Molecular Weight:
150.18
MDL number:
UNSPSC Code:
12352116
EC Index Number:
205-343-1
NACRES:
NA.22

Quality Level

form

solid

potency

65 mg/kg LD50, oral (Rat)

mp

84-87 °C

density

1.15 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H10N2O/c1-10(2)8-5-3-7(9-11)4-6-8/h3-6H,1-2H3

InChI key

CMEWLCATCRTSGF-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 82 °C
Melting range (upper value): ≤ 86 °C
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Self-heat. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service