Skip to Content
Merck
All Photos(1)

Key Documents

8.03105

Sigma-Aldrich

Diethyl sulfate

for synthesis

Synonym(s):

Diethyl sulfate, Sulfuric acid diethyl ester, Ethyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
154.18
MDL number:
EC Index Number:
200-589-6
Pricing and availability is not currently available.

vapor pressure

0.25 hPa ( 20 °C)

autoignition temp.

436 °C

potency

880 mg/kg LD50, oral (Rat)
944 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-12.2 % (v/v)

mp

-25 °C

transition temp

flash point 104 °C

solubility

7 g/L (Hydrolysis)

density

1.18 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H10O4S/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3

InChI key

DENRZWYUOJLTMF-UHFFFAOYSA-N

General description

for synthesis

Application

Diethyl sulfate for synthesis. CAS 64-67-5, chemical formula SO₂(OC₂H₅)₂.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(C)

104 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service