Skip to Content
Merck
All Photos(1)

Documents

8.02668

Sigma-Aldrich

Ethyl crotonate

for synthesis

Synonym(s):

Ethyl crotonate, Crotonic acid ethyl ester, trans-2-Butenoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOOC2H5
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352108
EC Index Number:
210-808-7
NACRES:
NA.22

vapor pressure

65 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

3000 mg/kg LD50, oral (Rat)

bp

138-140 °C/1013 hPa

transition temp

flash point 2 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H10O2/c1-3-5-6(7)8-4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

ZFDIRQKJPRINOQ-HWKANZROSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.916 - 0.918
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service