Skip to Content
Merck
All Photos(1)

Documents

8.02667

Sigma-Aldrich

Crotonaldehyde

(mixture of isomers) for synthesis

Synonym(s):

2-Butenal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.22

vapor pressure

24 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

230 °C

potency

80 mg/kg LD50, oral (rat)
128-324 mg/kg LD50, skin (rabbit)

expl. lim.

2.1-15.5 % (v/v)

bp

104 °C/1013 hPa

mp

-74 °C

transition temp

flash point 13 °C

solubility

181 g/L

density

0.85 g/cm3 at 20 °C

General description

mixture of isomers for synthesis

Application

Crotonaldehyde (mixture of isomers) for synthesis. CAS 4170-30-3, molar mass 70.09 g/mol.

Storage and Stability

Store below +30°C.

Analysis Note

Assay (GC, area%):≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C):0.851 - 0.853
Identity (IR):passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service