Skip to Content
Merck
All Photos(1)

Documents

681670

Sigma-Aldrich

Wogonin, S. baicalensis

A cell-permeable and orally available flavonoid that displays anti-inflammatory properties.

Synonym(s):

Wogonin, S. baicalensis, 5,7-Dihydroxy-8-methoxyflavone, WOG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C16H12O5/c1-20-15-12(19)7-10(17)14-11(18)8-13(21-16(14)15)9-5-3-2-4-6-9/h2-8,17,19H,1H3

InChI key

XLTFNNCXVBYBSX-UHFFFAOYSA-N

General description

A cell-permeable and orally available flavonoid that displays anti-inflammatory properties. Reported to suppress the release of NO by iNOS, PGE2 by COX-2, proinflammatory cytokines, and MCP-1 gene expression and NF-κB activation.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Lee, S.O., et al. 2006. Biochem. Biophys. Res. Comm.351, 118.
Kim, H.P., et al. 2004. J. Pharmacol. Sci.96, 229.
Chang, Y.L., et al. 2001. Mol. Pharmacol.60, 507.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service