65044B
Hemacolor® Solution II
Harleco® (Fixative for the rapid staining of blood smears, microorganisms and spermatozoa)
Synonym(s):
Hematology Stain
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
Quality Level
manufacturer/tradename
Harleco® (Fixative for the rapid staining of blood smears, microorganisms and spermatozoa)
mp
-0.1 °C
application(s)
diagnostic assay manufacturing
hematology
histology
shipped in
ambient
storage temp.
15-25°C
General description
HARLECO® Red color reagent for fast staining in hematology and cytology
Application
HARLECO® Red color reagent for fast staining in hematology and cytology
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
For In-Vitro Diagnostic Use.
For Professional Use.
Legal Information
HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany
HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service