Skip to Content
Merck
All Photos(1)

Documents

648493-M

Millipore

TRPML Agonist, ML-SA1

The TRPML Agonist, ML-SA1 controls the biological activity of TRPML. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

TRPML Agonist, ML-SA1, 2-(2-Oxo-2-(2,2,4-trimethyl-3,4-dihydroquinolin-1(2H)-yl)ethyl)isoindoline-1,3-dione, Mucolipin Synthetic Agonist 1, ML-SA1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N2O3
CAS Number:
Molecular Weight:
362.42
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

InChI

1S/C22H22N2O3/c1-14-12-22(2,3)24(18-11-7-6-8-15(14)18)19(25)13-23-20(26)16-9-4-5-10-17(16)21(23)27/h4-11,14H,12-13H2,1-3H3

InChI key

KDDHBJICVBONAX-UHFFFAOYSA-N

General description

A membrane permeable dihydroquinolinyloxo-isoindolinedione compound that acts as a potent and selective agonist of lysosomal mucolipin transient receptor potential (TRP) channel 1,2,3 (TRPML) in all mammalian cells. Does not appear to affect six other related TRP channels. Shown to rapidly activate whole-endolysosome TRPML-like current and induce lysosomal Ca2+ release (~10 µM). Its activating action on whole-endolysosome is comparable to that of PI(3,5)P2 (1 µM) with which it shows a synergistic effect. ML-SA1-induced Ca2+ responses are reportedly abolished by glycyl-L-phenylalanine 2-naphthylamide, induces osmotic lysis of lysosomes, and Bafilomycin A1 (>Cat. No. 196000), a V-ATPase inhibitor. Also reduces lactosylceramide and cholesterol accumulation and corrects trafficking defects in Niemann-Pick (NP) disease cells.

Biochem/physiol Actions

Primary Target
TRPML1,2,3

Warning

Toxicity: Standard Handling (A)

Other Notes

Shen, D., et al. 2012. Nat. Commun.3, 731.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service