Skip to Content
Merck
All Photos(1)

Key Documents

6.19117

Sigma-Aldrich

Vanillin

GR

Synonym(s):

Vanillin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)(OH)C6H3CHO
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352100

SMILES string

O(C)c1c(ccc(c1)C=O)O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3

InChI key

MWOOGOJBHIARFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service