Skip to Content
Merck
All Photos(1)

Key Documents

512731

Millipore

Paromomycin Sulfate

An aminoglycoside antibiotic containing 5 amino groups that exhibits antibacterial and antiamebic activity.

Synonym(s):

Paromomycin Sulfate, Neomycin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H45N5O14 · xH2SO4
CAS Number:
Molecular Weight:
615.63 (free base basis)
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

water: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C23H45N5O14.H2O4S/c24-2-7-13(32)15(34)10(27)21(37-7)41-19-9(4-30)39-23(17(19)36)42-20-12(31)5(25)1-6(26)18(20)40-22-11(28)16(35)14(33)8(3-29)38-22;1-5(2,3)4/h5-23,29-36H,1-4,24-28H2;(H2,1,2,3,4)/t5-,6+,7+,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18?,19-,20-,21-,22-,23?;/m1./s1

InChI key

LJRDOKAZOAKLDU-UMIPPVEZSA-N

General description

An aminoglycoside antibiotic containing 5 amino groups that exhibits antibacterial and antiamebic activity. Recent NMR evidence indicates that paromomycin binding to the A-site of 16S rRNA induces a local conformational change, suggesting a mechanism for aminoglycoside action on protein translation. Also exhibits anti-protozoal properties.
Potency: ≥675 µg/mg.

Warning

Toxicity: Harmful (C)

Other Notes

VanLoock, M.S., et al. 1999. J. Mol. Biol. 285, 2069.
Blanchard, S.C., et al. 1998. Biochemistry 37, 7716.
Fourmy, D., et al. 1998. J. Mol. Biol. 277, 333.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Shahiduzzaman et al.
Experimental parasitology, 212, 107873-107873 (2020-03-14)
Ginsenoside-Rh2 and cucurbitacin-B (CuB) are secondary metabolites of Ginseng (Panax ginseng) and Cucurbitaceae plants respectively. We assessed the anticryptosporidial activity of these two functional compounds in a cell culture model of cryptosporidiosis. The highest concentration of each compound that was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service