Skip to Content
Merck
All Photos(1)

Key Documents

5.09721

Millipore

1α,25-Dihydroxy Vitamin D3

InSolution, ≥99%

Synonym(s):

InSolution Vitamin D3, 1α, 25-Dihydroxy-, 1,25-(OH)₂-D₃

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
Molecular Weight:
416.64
UNSPSC Code:
12352205
NACRES:
NA.21

Quality Level

Assay

≥99% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

General description

Active hormonal form of vitamin D, formed by the renal 1-hydroxylation of 25-(OH)D3. Has immunomodulatory properties in vitro and in vivo. Prevents the development of clinical diabetes in NOD mice, which are an animal model of human autoimmune diabetes. Induces reversible hydrolysis of the sphingomyelin cycle and generates ceramide. Stimulates intestinal Ca2+ and phosphate transport and mobilizes Ca2+ from bone. Involved in cellular differentiation, carcinogenesis, and insulin secretion.

Application


  • Evaluation system for cell-permeable CYP3A4 inhibitory activity using 1α,25-dihydroxy-vitamin D(3)-induced intestinal cell lines.: Ueno et al. developed an evaluation system for cell-permeable CYP3A4 inhibitory activity utilizing 1α,25-dihydroxy-vitamin D3-induced intestinal cell lines. This system helps in screening for CYP3A4 inhibitors, which are critical in drug metabolism and interaction studies (Ueno et al., 2021).

Biochem/physiol Actions

Primary Target
vitamin D receptor

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 1 mM (50 ug/112 µL) solution of Vitamin D3, 1α, 25-Dihydroxy- (Cat. No. 679101) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Semizarov, D., et al. 1998. Proc. Natl. Acad. Sci. USA95, 15412.
Hannun, Y.A. 1994. J. Biol. Chem.269, 3125.
Mathieu, C., et al. 1994. Diabetologica37, 552.
Thomasset, M. 1994. Pathol. Biol.42, 163.
Horton, W.E., Jr., et al. 1991. J. Biol. Chem.266, 24804.
Larsen, C.G., et al. 1991. Biochem. Biophys. Res. Commun.176, 1020.
Thavarajah, M., et al. 1991. Biochem. Biophys. Res. Commun.176, 1189.
Wakasugi, M., et al. 1991. Prostaglandins42, 127.
Matsumoto, K., et al. 1990. Biochem. Biophys. Res. Commun.166, 916.
Simpson, R.U., et al. 1989. J. Biol. Chem.264, 19710.
DeLuca, H.F., and Schnoes, H.K. 1983. Annu. Rev. Biochem.52, 411.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service