Skip to Content
Merck
All Photos(1)

Key Documents

4.90061

Sigma-Aldrich

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

for synthesis

Synonym(s):

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11F3N2O3S
CAS Number:
Molecular Weight:
260.23
MDL number:
UNSPSC Code:
12352107

Quality Level

Assay

≥98.0% (HPLC)

form

liquid

mp

-12 °C

density

1.386 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H11N2.CHF3O3S/c1-3-8-5-4-7(2)6-8;2-1(3,4)8(5,6)7/h4-6H,3H2,1-2H3;(H,5,6,7)/q+1;/p-1

InChI key

ZPTRYWVRCNOTAS-UHFFFAOYSA-M

Analysis Note

Purity (HPLC): ≥ 98.0 %
Identity (NMR): passes test
Halides (IC): ≤ 0.1 %
Water (K. F.): ≤ 1.0 %

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

425.3 °F - closed cup

Flash Point(C)

218.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service