Skip to Content
Merck
All Photos(1)

Key Documents

245-M

Sigma-Aldrich

Indigo Carmine

HARLECO® For staining cytoplasm, negri bodies and connective tissue

Synonym(s):

Indigo Carmine, Carmine Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2Na2O8S2
CAS Number:
Molecular Weight:
466.35
Colour Index Number:
73015
MDL number:
UNSPSC Code:
41116130
EC Index Number:
212-728-8

Quality Level

pH

7 (20 °C, 10 g/L in H2O)

mp

>340 °C (decomposition)

solubility

1 g/L

storage temp.

10-30°C

InChI

1S/C16H10N2O8S2.2Na/c19-15-9-5-7(27(21,22)23)1-3-11(9)17-13(15)14-16(20)10-6-8(28(24,25)26)2-4-12(10)18-14;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;

InChI key

KHLVKKOJDHCJMG-QDBORUFSSA-L

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used for staining cytoplasm, negri bodies and connective tissue. Can be mixed with Eosin Y as a counterstain to hematoxylin for staining vaginal smears.
HARLECO® For staining cytoplasm, negri bodies and connective tissue

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service