Skip to Content
Merck
All Photos(1)

Key Documents

217697-M

Sigma-Aldrich

Cdk1 Inhibitor III

The Cdk1 Inhibitor III, also referenced under CAS 440362-74-3, controls the biological activity of Cdk1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk1 Inhibitor III, Ethyl-(6-hydroxy-4-phenylbenzo[4,5]furo[2,3-b])pyridine-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15NO4
CAS Number:
Molecular Weight:
333.34
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

powder

potency

28.8 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

ethanol: 10 mg/mL
DMSO: 20 mg/mL

storage temp.

2-8°C

General description

This product has been discontinued.



A cell-permeable 1-aza-9-oxafluorene compound that acts as a highly selective Cdk1/cyclin B inhibitor (IC50 = 28.8 µM) with very little activity against Cdk5/p25, Cdk2/E, or Cdk4/D1. Unlike other Cdk inhibitors, it exhibits potent inhibitory effect against P-glycoprotein-mediated MDR (multidrug resistance) in cancer cells (~9-fold more potent than verapamil). Its special bifunctional cytostatic property makes it an excellent tool for research in cancer treatment.

A cell-permeable, highly selective Cdk1/cyclin B inhibitor (IC50 = 28.8 µM). Exhibits only low inhibitory activity against Cdk5/p25, Cdk2/cyclin E, and Cdk4/cyclin D1. Unlike other Cdk inhibitors has also been reported to have potent inhibitory effects on P-glycoprotein-mediated multidrug resistance (MDR) in cancer cells (~9-fold more potent than verapamil). Its unique bifunctional cytostatic properties make it an attractive tool in cancer research.

Biochem/physiol Actions

Primary Target
Cdk1/cyclin B

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Brachwitz, K., et al. 2003. J. Med. Chem.46, 876.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service