Skip to Content
Merck
All Photos(1)

Documents

20-218

Millipore

Ultrafiltration Spin-Columns, 0.45µm cutoff

Spin-Columns, each consisting of a 750μl capacity filter insert

Synonym(s):

Centrifuge filter tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.24

Quality Level

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

shipped in

ambient

Quality

For use to clear cell lysates prior to immunoprecipitation, ensuring a cleaner and more reproducible result. Ultrafiltration Spin-Columns are especially useful in situations where old or frozen lysates are used, or when cell debris has not been adequately cleared during cell lysis.

Storage and Stability

5 years at room temperature

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service