Skip to Content
Merck
All Photos(1)

Key Documents

1.50226

Supelco

MANU-CART 55 mm HPLC cartridge holder

for LiChroCART 55-2, 55-3 and 55-4 mm HPLC-cartridges

Synonym(s):

TM=["MANU-CART"] holder for TM=["LICHROCART"] HPLC-cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

manu-CART 55 mm cartridge holder for LiChroCART 55-2, 55-3 and 55-4 mm HPLC-cartridges,

technique(s)

HPLC: suitable

Quality Level

storage temp.

no temp limit

General description

manu-CART holder 55 mm for 55-2, 55-3 and 55-4 LiChroCART cartridges.

Analysis Note

visual control: passes test
requirement of the german Zusatzstoff-Verkehrsverordnung.

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service