Skip to Content
Merck

1.37060

SAFC

Sodium hydroxide solution

EMPROVE® EXPERT, suitable for cleaning in place, 0,5 mol/L

Pharma Manufacturing

Synonym(s):

Sodium hydroxide solution 0,5 mol/L

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305

vapor pressure

3 mmHg ( 37 °C)

Quality Level

product line

EMPROVE® EXPERT

form

liquid

concentration

≤0.2% Na2CO3 (Carbonate)
≤10 ppm Al (Aluminium)
≤3 ppm Fe
≤30 ppm Pb (Heavy metals)
≤30 ppm sulfate (SO<SUB>4</SUB>)
≤5 ppm chloride (CL)
0.45-0.55 mmol/L NaOH (acidimetric)

impurities

≤2.0 IU/mL Endotoxins

pH

13.5 (20 °C, H2O)

conductivity

80-100 mS/cm at 22 °C

density

1.02 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Components

This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and water for injection USP. Water for injection USP has been prepared for this product by using purified water according to Ph Eur, JP.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service