Skip to Content
Merck
All Photos(1)

Documents

1.32225

Supelco

Aluminium Standard Solution

traceable to SRM from NIST 0.200 mg/l Al in H2O

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

Quality Level

form

liquid

specific analyte(s)

aluminium

technique(s)

photometry: suitable

pH

3-5 (20 °C in H2O)

density

1 g/cm3

storage temp.

15-25°C

General description

This Spectroquant® Aluminium Standard Solution is a high-quality laboratory solution used for internal quality control and method controlling purposes.

These Spectroquant® standard solutions perfectly complement our ready-to-use test kits and photometers within the Spectroquant® portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.

Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.

Application

Aluminium Standard Solution can be used to check the photometric measurement system (reagents, measurement device, handling) and the mode of working.

Features and Benefits

  • Manufactured and tested in DIN EN ISO 9001:2008 accredited laboratory
  • The solution is pre-diluted to a concentration that falls into the range of our Spectroquant® test kits. This eliminates the need for dilution prior to analysis and hence avoids a common source of error
  • Ready-to-use diluted reference materials (RMs) along with expanded measurement uncertainty ensure absolute precision
  • Accompanied with a certificate of analysis (COA) detailing the certified values, including uncertainty and expiry date
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST) or to United States Pharmacopeia (USP)
  • Reliable and reproducible results

Analysis Note

Concentration: 0.200 ± 0.006 mg/L Al in H2O

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination with Chromazurole S subsequent to fusion melting

Photometric determination with Chromazurole S subsequent to fusion melting

Photometric determination with Chromazurole S subsequent to fusion melting

Photometric determination with Chromazurole S subsequent to fusion melting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service