Skip to Content
Merck
All Photos(1)

Documents

121765

Sigma-Aldrich

AG 825

A potent, cell-premeable, reversible, substrate competitive, and selective inhibitor of HER2 (neu/ErbB2; IC₅₀ = 0.35 µM) relative to HER1 (IC₅₀= 19 µM) autophosphorylation.

Synonym(s):

AG 825, 4-Hydroxy-3-methoxy-5-(benzothiazolylthiomethyl)benzylidenecyanoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15N3O3S2
CAS Number:
Molecular Weight:
397.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C19H15N3O3S2/c1-25-15-8-11(6-12(9-20)18(21)24)7-13(17(15)23)10-26-19-22-14-4-2-3-5-16(14)27-19/h2-8,23H,10H2,1H3,(H2,21,24)/b12-6+

InChI key

KXDONFLNGBQLTN-WUXMJOGZSA-N

General description

A potent, cell-premeable, reversible, substrate competitive, and selective inhibitor of HER-2 (neu/erbB-2, IC50 = 0.35 µM) relative to HER-1 (IC50 = 19 µM) autophosphorylation. The inhibition is competitive with respect to ATP binding. Enhances the chemosensitivity in non-small cell lung cancer (NSCLC) cell lines expressing high levels of p185neu.
A potent, cell-premeable, reversible, substrate competitive, and selective inhibitor of HER2 (neu/ErbB2; IC50 = 0.35 µM) relative to HER1 (IC50 = 19 µM) autophosphorylation. The inhibition is competitive with respect to ATP binding. Enhances chemosensitivity in non-small cell lung cancer (NSCLC) cell lines expressing high levels of p185neu.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HER2
Product does not compete with ATP.
Reversible: yes
Target IC50: 0.35 µM against HER2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2-3 months at -20°C.

Other Notes

Ashton, J.M., et al. 2012. Cell Stem Cell11, 359.
Tsai, C.M., et al. 1996. Cancer Res.56, 1068.
Levitzki, A., and Gazit, A. 1995. Science267, 1782.
Osherov, N., et al. 1993. J. Biol. Chem. 268, 11134.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service