1.14780
Zinc Test Kit, colorimetric
0.1-5 mg/L (Zn), MQuant®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Zinc Test, colorimetric, 0.1-5 mg/L (Zn), for use with MQUANT®
specific analyte(s)
zinc
Quality Level
measuring range
0.1-5 mg/L (Zn)
compatibility
for use with MQUANT®
detection method
colorimetric
storage temp.
15-25°C
General description
Method: colorimetric with color-disk comparator 0.1 - 0.2 - 0.3 - 0.4 - 0.5 - 0.7 - 1 - 2 - 5 mg/l Zn MQuant®
The MQuant® Zinc test (formally MColortest®) measures zinc ions in:
Method
In sulfuric solution zinc ions react with thiocyanate ions and brilliant green to form a blue-green complex. The zinc concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.14782 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.
- Groundwater and surface water
- Drinking water and mineral water
- Beverages
- Industrial water
- Boiler water
- Wastewater and percolating water
- Electroplating wastewater
Method
In sulfuric solution zinc ions react with thiocyanate ions and brilliant green to form a blue-green complex. The zinc concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.14782 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.
Legal Information
MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 2 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service