Skip to Content
Merck
All Photos(1)

Key Documents

1.11969

Sigma-Aldrich

Acetone-D6

deuteration degree min. 99.96% for NMR spectroscopy MagniSolv

Synonym(s):

Acetone-D6, Hexadeuteroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3D6O
CAS Number:
Molecular Weight:
64.12
MDL number:
UNSPSC Code:
12142201
EC Index Number:
211-563-9

vapor pressure

233 hPa ( 20 °C)

form

liquid

autoignition temp.

465 °C (DIN 51794)

potency

5800 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.6-12.8 % (v/v)

technique(s)

NMR: suitable

impurities

≤0.010% Water (H<SUB>2</SUB>O) (NMR)
≤0.020% Water (H<SUB>2</SUB>O + D<SUB>2</SUB>O) (Karl Fischer)

pH

5-6 (365 g/L in H2O)

bp

56.2 °C/1013 hPa

mp

-95.4 °C

transition temp

flash point <-20 °C

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3

InChI key

CSCPPACGZOOCGX-WFGJKAKNSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Deuteration degree: ≥ 99.96 %
Water (NMR, H₂O): ≤ 0.010 %
Performance test (NMR-spectrum): passes test
Water (K. Fischer, H₂O + D₂O): ≤ 0.020 %

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(C)

-19 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service