Skip to Content
Merck
All Photos(1)

Key Documents

1.09754

Supelco

Methyl margarate

reference substance for gas chromatography

Synonym(s):

Methyl margarate, Methyl heptadecanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36O2
CAS Number:
Molecular Weight:
284.48
MDL number:
UNSPSC Code:
12352108
EC Index Number:
217-055-3

Quality Level

form

solid

bp

184 °C/13.3 hPa

mp

28-30 °C

transition temp

flash point >110 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3

InChI key

HUEBIMLTDXKIPR-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99.0
Identity (IR-spectrum): conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

> 110.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service