Skip to Content
Merck
All Photos(1)

Key Documents

1.08712

Supelco

Magnesium reagent

according to Mann and Yoe as reagent for magnesium

Synonym(s):

Magnesium reagent, Xylidyl blue, Mann and Yoe′s magnesium reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20N3NaO6S
CAS Number:
Molecular Weight:
513.50
MDL number:
UNSPSC Code:
12352107
EC Index Number:
239-012-8

form

solid

Quality Level

feature

according to Mann and Yoe as reagent for magnesium

specific analyte(s)

magnesium

technique(s)

photometry: suitable

storage temp.

15-25°C

InChI

1S/C25H21N3O6S.Na/c1-14-7-9-20(15(2)11-14)26-25(31)19-12-16-5-3-4-6-18(16)23(24(19)30)28-27-21-13-17(35(32,33)34)8-10-22(21)29;/h3-13,27,29H,1-2H3,(H,26,31)(H,32,33,34);/q;+1/p-1/b28-23-;

InChI key

LPXSXFXFNSMAOI-MCOPDZHSSA-M

General description

acc. to Mann and Yoe (reagent for magnesium)

Analysis Note

Identity (IR-spectrum): passes test
Absorbance A (492 nm; 1 cm): ≥ 0.350
Loss on Drying (105°C): ≤ 5

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service