About This Item
Recommended Products
form
solid
Quality Level
composition
Dye, ≥90% spectrophotometric assay
pH
5.5-7.0 (20 °C, 1 g/L in H2O)
solubility
1 g/L
bulk density
400‑450 kg/m3
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
InChI
1S/C14H9NO7S.Na/c15-9-7-8(12(18)14(13(9)19)23(20,21)22)11(17)6-4-2-1-3-5(6)10(7)16;/h1-4,18-19H,15H2,(H,20,21,22);/q;+1/p-1
InChI key
IFSXZLJQEKGQAF-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Analysis Note
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ 1 (Dimethyl sulfoxide): 557 - 559 nm
Absorption maximum λ 2 (Dimethyl sulfoxide): 520 - 522 nm
Spec. Absorptivity A 1%/1cm
- λ1max; 0.02 g/l; dimethyl sulphoxide:
380 - 422
- λλ2max; 0.02 g/l; Dimethyl sulphoxide: 383 - 426
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for microscopy: passes test
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service