1.02931
Dimethyl sulfoxide
for DNA and peptide synthesis (max. 0.025% H₂O)
Synonym(s):
Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO, DMSO
About This Item
Recommended Products
vapor density
2.7 (vs air)
Quality Level
vapor pressure
0.42 mmHg ( 20 °C)
Assay
≥99.9% (GC)
form
liquid
autoignition temp.
573 °F
potency
28300 mg/kg LD50, oral (Rat)
40000 mg/kg LD50, skin (Rat)
expl. lim.
42 %, 63 °F
technique(s)
DNA amplification: suitable
impurities
≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.025% Water
evapn. residue
≤10 mg/L
color
APHA: ≤10
refractive index
n20/D 1.479 (lit.)
kinematic viscosity
2.14 cSt
bp
189 °C (lit.)
mp
16-19 °C (lit.)
transition temp
flash point 87 °C
solubility
1000 g/L
density
1.10 g/mL (lit.)
application(s)
peptide synthesis
storage temp.
2-30°C
SMILES string
CS(C)=O
InChI
1S/C2H6OS/c1-4(2)3/h1-2H3
InChI key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Dimethyl sulfoxide as a gas phase charge-reducing agent for the determination of PEGylated proteins′ intact mass.: This study explores the effectiveness of DMSO in reducing the charge of proteins during mass spectrometry, improving the accuracy of mass measurements for PEGylated proteins, a critical aspect in biopharmaceutical analyses (Skjærvø Ø et al., 2024).
- Benzoselenadiazole-Functionalized H-Bonded Arylamide Foldamers: Solvent-Responsive Properties and Helix Self-Assembly Directed by Chalcogen Bonding in Solid State.: This paper discusses the role of DMSO in influencing the self-assembly properties of novel foldamers, showcasing its importance in the study of advanced material sciences (Liu CZ et al., 2024).
- Controlling Interfacial Hydrogen Bonding at a Gold Surface: The Effect of Organic Cosolvents.: This study presents DMSO′s role in modulating hydrogen bonding at interfaces, particularly in the presence of gold surfaces, which is relevant in surface chemistry and sensor applications (Al-Mualem ZA et al., 2024).
Caution
Specifications
Analysis Note
Identity (IR): conforms
Color: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.025 %
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service