1.02924
DAB-buffer tablets
suitable for immunohistochemistry
Synonym(s):
3,3-Diamino benzidine
About This Item
Recommended Products
form
tablet
Quality Level
IVD
for in vitro diagnostic use
technique(s)
immunohistochemistry: suitable
color
beige
pH
6.6 (25 °C, 25 g/L in H2O)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-8°C
Related Categories
General description
The package is sufficient for 50 applications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B - Muta. 2
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service