Skip to Content
Merck
All Photos(1)

Documents

1.02242

Sigma-Aldrich

Casein acc. to HAMMARSTEN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.21

form

solid

solubility

20.1 g/L (experimental)

storage temp.

20-25°C

Linkage

CX0525

Analysis Note

Assay (ex N, calc. on dried substance):≥ 95 %
Free acid (as lactic acid):≤ 0.1 %
In water soluble matters:≤ 0.2 %
Fats:≤ 0.1 %
Sulfated ash (800 °C):≤ 1.5 %
Loss on Drying (105 °C):≤ 6 %
Microbiological test:passes test
Lactose (HPLC):≤ 0.5 %
Glucose (HPLC):≤ 0.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service