Skip to Content
Merck
All Photos(1)

Key Documents

04-959

Sigma-Aldrich

Anti-Retinoid-Related Orphan Receptor γ T Antibody, clone RORg2

clone RORg2, from hamster(Armenian)

Synonym(s):

RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, nuclear receptor ROR-gamma, retinoid-related orphan receptor gamma, Nuclear receptor RZR-gamma, RZR-GAMMA, Nuclear receptor subfamily 1 group F member

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

hamster (Armenian)

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RORg2, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rorc(19885)

General description

Retinoid-related orphan receptor-gamma t (RORγT) is one of two RORγ isoforms belonging to the nuclear hormone receptor family and contains a single nuclear receptor DNA-binding domain. RORγT is a negative regulator of TNFDF6/FASL expression and T-cell production of IL2. The upregulation of RORγT is a requirement for TH17 cell differentiation. Inhibition of RORγT is induced by TGF-β’s interaction with Foxp3. Expression is observed mainly in immature thymocytes, and, during fetal development, is found exclusively in lymphoid-inducer (LTi) cells.

Immunogen

Epitope: Unknown
Histidine-tagged recombinant protein corresponding to mouse Retinoid-Related Orphan Receptor γ T (RORγT).

Application

Detect Retinoid-Related Orphan Receptor γ T using this Anti-Retinoid-Related Orphan Receptor γ T Antibody, clone RORg2 validated for use in WB, FC & IF.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Quality

Evaluated by Western Blot in EL4 cell lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected Retinoid-Related Orphan Receptor γ T (RORγT) in 10 µg of EL4 cell lysate.

Target description

~52 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified hamster monoclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
EL4 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Sheng Bi et al.
Cell proliferation, 53(6), e12827-e12827 (2020-05-15)
Previously, we found that by regulating T helper (Th) cell polarization, calcitriol intervention inhibited lipopolysaccharide (LPS)-induced alveolar bone loss in an animal periodontitis model, but the underlying cellular events remain unknown. In this study, mouse Th cells were incubated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service