04-893
Anti-JNK3/SAPK1b Antibody, clone C05T, rabbit monoclonal
culture supernatant, clone C05T, from rabbit
Synonym(s):
JNK3 alpha protein kinase, MAP kinase, MAP kinase p49 3F12, Stress-activated protein kinase JNK3, c-Jun N-terminal kinase 3, c-Jun kinase 3, mitogen-activated protein kinase 10, stress activated protein kinase JNK3, stress activated protein kinase beta
About This Item
biological source
rabbit
Quality Level
antibody form
culture supernatant
antibody product type
primary antibodies
clone
C05T, monoclonal
species reactivity
rat, human
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... MAPK10(5602)
rat ... Mapk10(25272)
General description
The JNK/SAPK1 kinases, like the other MAPK-like kinases, are thought to phosphorylate multiple substrates and regulate many processes, including proliferation (in some cell types) and apoptosis. The SAPK2/3 family is most widely referred to as the p38 family. These kinases are also activated by stresses, most notably inflammatory cytokines, irradiation, and certain toxins such as anisomycin and arsenite. The activating kinases of SAPK2/3 are SKK2/MEK3 for SAPK2a and 2b, and MKK6 for SAPK3. The targets of the SAPK2/3 family include the MAPKAP kinases 2 and 3/3pK. In addition, SKK4 is related to this family, exhibiting 60% identity, and is activated by MKK6.
Specificity
Immunogen
Application
Signaling
Kinases & Phosphatases
Quality
Target description
Linkage
Physical form
Storage and Stability
Analysis Note
RIPA cell lysates from A431 cells.
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service