Skip to Content
Merck
All Photos(1)

Documents

KEY001126156

Key Organics / BIONET

1-(6-chloropyridazin-3-yl)piperidine-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
240.69
MDL number:

mp

216 - 218

Iupac Name

1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxamide

Inchi Code

1S/C10H13ClN4O/c11-8-1-2-9(14-13-8)15-5-3-7(4-6-15)10(12)16/h1-2,7H,3-6H2,(H2,12,16)

InChI key

BTUDQZJQBUPWRV-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/11F-905

Melting Point

216 - 218

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/11F-905


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service