Key Documents
Z764558
2mag bioMIXcontrol S control unit
AC/DC input 110 V AC, US 3-pin plug
Synonym(s):
induction stirrer, magnetic induction stir plate, magnetic wear-free stirrer, stirrer
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
2mag 98500
parameter
5-250 rpm speed
W × D × H
215 mm × 120 mm × 55 mm
plate size × W × L
× 200 mm × 250 mm
weight
1.4 kg
output
4-40 W (stirring power)
General description
  • Efficient control unit, for 2mag bioMIXdrive series, developed for the hard daily lab use and uncompromising warming-free continuous operation
  • Control unit for the connection of 1 stirring drive bioMIXdrive 1, 2, 3, 4, or 6 with 6 watts/stirring point, without any own-generated heating effect, extended speed range from 5 to 250 rpm, jerk-free stirring, even and smooth stirring also at slow speeds
  • Power setting (10-step) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation, e.g. in CO2-incubators
  • Identical to bioMIXcontrol, but with stackable housing and vertical control panel
  • Digital display for speed- and power setting, SoftStart for reliable and safe accelerating for stirring bar, QuickSet for quick setting of start- and maximum speed, storage of last working status when turning off, firm stainless steel housing, and easy to clean
required but not provided
Choose from one of the most recent versions:
Certificates of Analysis (COA)