Skip to Content
Merck
All Photos(1)

Key Documents

Z423270

Best® N-DEX® laboratory gloves

powder-free, thickness 4 mil (0.12 mm), size 8‑9 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504
NACRES:
NB.35

material

blue
nitrile

size

8‑9 L

sterility

non-sterile

feature

disposable
latex free
powder-free

packaging

pack of 100 ea (boxes of 100)

manufacturer/tradename

Best Manufacturing 7005PFL

L

240 mm

thickness

4 mil (0.12 mm)

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Soft elastic material feels cooler than natural rubber or latex and conforms to hand for greater comfort and dexterity.

  • 100% nitrile non-latex glove
  • Ambidextrous
  • Excellent alternative to latex
  • Rolled cuff

Legal Information

Best is a registered trademark of Showa Glove Co.
N-DEX is a registered trademark of Showa Glove Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service