Key Documents
Z313874
Retsch test sieves
2.8 mm, D × H 200 mm × 50 mm
About This Item
UNSPSC Code:
41105003
material
stainless steel (with wire gauze)
manufacturer/tradename
Retsch
availability
not available in USA and Canada
D × H
200 mm × 50 mm
particle size
2.8 mm
General description
For mechanical or manual, wet or dry sieving. Quintuple tested and provided with Quality Certificate DIN ISO 3310/1.
Choose from one of the most recent versions:
Certificates of Analysis (COA)