Z263370
Wheaton straight-mouth bottles with cap, clear
capacity 4 oz (125 mL), white polypropylene cap, vinyl flat liner
Synonym(s):
Wheaton bottles, bottles clear, clear bottles
About This Item
Recommended Products
material
round bottle
soda-lime glass bottle
vinyl flat liner
white polypropylene cap
cap size
58-400
Joint
Joints threaded neck (58-400)
sterility
non-sterile
feature
graduations: no
with cap
packaging
pkg of 24 ea
manufacturer/tradename
WHEATON® W216904
capacity
4 oz (125 mL)
diam. × H
60 mm × 68 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service