Key Documents
Z197904
Flexible bonding wire
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Pricing and availability is not currently available.
manufacturer/tradename
Justrite 8499
L
0.91 m
3 ft
size
General description
Prevents sparking of static electricity generated by the transfer of flammable liquids.
Choose from one of the most recent versions:
Certificates of Analysis (COA)