Z153885
Duran® plastic-coated laboratory bottles
capacity 250 mL
Synonym(s):
bottle laboratory, lab bottle
About This Item
Recommended Products
material
borosilicate glass 3.3
clear bottle
narrow-mouth bottle
round bottle
feature
autoclavable
without cap
packaging
pack of 1 ea
pack of 10 ea
manufacturer/tradename
DURAN® 21 805 36
parameter
max. pressure (Not for use under pressure)
capacity
250 mL
diam. × H
70 mm × 138 mm
fitting
GL 45 neck thread
Looking for similar products? Visit Product Comparison Guide
General description
Please note: Cap not included.
Features and Benefits
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- The external polymer coating provides scratch, leak* and splinter* protection (* only applies to bottles 2000 ml and less)
- Highly resistant, long service-life coating
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- UV protection up to approx. 380 nm due to external coating
- Complete with screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
- Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Legal Information
required but not provided
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service