Key Documents
Z112488
Sigma-Aldrich
Magnetic holder
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
technique(s)
IR spectroscopy: suitable
General description
  • Holds standard 13 mm KBr pellets and smaller pellets with paper inserts
  • Holds crystal windows listed (requires magnetic strips)
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)