Skip to Content
Merck
All Photos(1)

Key Documents

W510319

Sigma-Aldrich

Methyl vanillate

≥98%, FG

Synonym(s):

Methyl 4-hydroxy-3-methoxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CO2CH3
CAS Number:
Molecular Weight:
182.17
EC Number:
Council of Europe no.:
2305
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.799
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥98%

bp

285-287 °C (lit.)

mp

64-67 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

vanilla

SMILES string

COC(=O)c1ccc(O)c(OC)c1

InChI

1S/C9H10O4/c1-12-8-5-6(9(11)13-2)3-4-7(8)10/h3-5,10H,1-2H3

InChI key

BVWTXUYLKBHMOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl vanillate (MV) occurs naturally in vanilla beans and Pinot noir wine. It is one of the degradation products of Kraft lignin. In vitro studies in mice indicates that MV may be the potent ingredient of Hovenia dulcis Thunb (HDT) extract that is responsible for its ability to increase bone mass, which makes it a potential lead as an anabolic agent for the development of anti-osteoporosis drugs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hovenia dulcis Thunb extract and its ingredient methyl vanillate activate Wnt/?-catenin pathway and increase bone mass in growing or ovariectomized mice.
Cha PH, et al.
PLoS ONE, 9(1), e85546-e85546 (2014)
Headspace Techniques Utilized for the Detection of Volatile Flavor Compounds of the Vanilla Bean.
Klimes I & Lamparsky D.
Analysis of Foods and Beverages, 95-114 (1978)
Oxidation of lignin using aqueous polyoxometalates in the presence of alcohols.
Voitl T & Rudolf von Rohr P.
ChemSusChem, 1(8?9), 763-769 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service