Skip to Content
Merck
All Photos(1)

Documents

W372102

Sigma-Aldrich

2,4-Octadienal, predominantly trans,trans

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH=CHCHO
CAS Number:
Molecular Weight:
124.18
FEMA Number:
3721
Council of Europe no.:
11805
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.186

biological source

synthetic

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥95%

composition

trans,trans isomer, ≥85%

refractive index

n20/D 1.526 (lit.)

bp

88 °C/10 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; melon

storage temp.

2-8°C

SMILES string

CCC\C=C\C=C\C=O

InChI

1S/C8H12O/c1-2-3-4-5-6-7-8-9/h4-8H,2-3H2,1H3/b5-4+,7-6+

InChI key

DVVATNQISMINCX-YTXTXJHMSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuliana D'Ippolito et al.
Organic letters, 5(6), 885-887 (2003-03-14)
[reaction: see text] Marine diatoms produce alpha,beta,gamma,delta-unsaturated aldehydes that have detrimental effects on the reproduction of their natural predators. The production of these defensive metabolites is suggested to involve enzymatic oxidation of polyunsaturated fatty acids. In this paper, feeding experiments
Francois Ribalet et al.
Aquatic toxicology (Amsterdam, Netherlands), 86(2), 249-255 (2007-12-21)
Bioactive polyunsaturated aldehydes (PUAs) are produced by several marine phytoplankton (mainly diatoms) and have been shown to have a detrimental effect on a wide variety of organisms, including phytoplankton and invertebrates. However, their potential impact on marine bacteria has been
Christian Galasso et al.
International journal of molecular sciences, 21(15) (2020-07-28)
Programmed cell death, such as apoptosis and autophagy, are key processes that are activated early on during development, leading to remodelling in embryos and homeostasis in adult organisms. Genomic conservation of death factors has been largely investigated in the animal
Giuliana d'Ippolito et al.
Phytochemistry, 67(3), 314-322 (2006-01-03)
Intermediates of the aldehyde biosynthesis in Thalassiosira rotula are investigated. Use of labeled precursors and cell preparations proves production of 2E,4Z-octadienal from 6Z,9Z,12Z-hexadecatrienoic acid (C16:3 omega-4) through the lipoxygenase-dependent intermediate (9S)-9-hydroperoxyhexadeca-6,10,12-trienoic acid. On the contrary, synthesis of 2E,4Z,7Z-decatrienal involves mainly
Roberta Esposito et al.
International journal of molecular sciences, 21(3) (2020-01-26)
Oxylipins are diatom-derived secondary metabolites, deriving from the oxidation of polyunsatured fatty acids that are released from cell membranes after cell damage or senescence of these single-celled algae. Previous results revealed harmful toxic effects of polyunsaturated aldehydes (PUAs) and hydroxyacids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service